Anti-TLR 11 Rabbit Polyclonal Antibody
Catalog # BIRBORB108995-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:TLR 11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Rabbit polyclonal TLR11 antibody was raised against a peptide corresponding to 15 amino acids near the carboxy-terminus of mouse TLR11.
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TLR 11
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse