Anti-LPP1/1A Rabbit Polyclonal Antibody
BIRBORB108864-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:LPP1/1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide derived from the LPP1 protein
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: LPP1/1A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human