Anti-Keratin 13 + 10 Mouse Monoclonal Antibody
Catalog # BIRBORB108791-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Keratin 13/10
- Antigen symbol:KRT13/10
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Hybridoma produced by the fusion of splenocytes from mice immunized with cytoskeleton preparation from HeLa cells and mouse myeloma cells.
- Purification:purified by Protein A/G Chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Keratin 13 + 10
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human