Anti-BIScV Rabbit Polyclonal Antibody
BIRBORB48711-200
: Biorbyt
- Antibody type:Primary
- Antigen symbol:BISCV
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Format:ascites fluid
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Peptide
- Purification:purified by affinity chromatography
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: BIScV
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Virus