Anti-SLC22A5 Rabbit Polyclonal Antibody
BIRBORB48540-100
: Biorbyt
- Antibody type:Primary
- Antigen name:solute carrier family 22 (organic cation/carnitine transporter), member 5
- Antigen symbol:Solute carrier family 22, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SLC22A5 (solute carrier family 22 (organic cation/carnitine transporter), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat