Anti-TCR alpha Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB44731-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:T cell receptor alpha chain
- Antigen symbol:TCRA
- Clonality:Monoclonal
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen synonyms:TCR alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TCR alpha
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human