Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD7 Mouse Monoclonal Antibody
Anti-CD7 Mouse Monoclonal Antibody
Catalog # BIRBORB44608-100
Supplier:  Biorbyt
Anti-CD7 Mouse Monoclonal Antibody
Catalog # BIRBORB44608-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD7 molecule
  • Antigen symbol:
    CD7
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 test
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: CD7 (CD7 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human