Anti-ITGAL Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB44220-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Integrin, Alpha L (Antigen CD11A (P180), Lymphocyte Function-Associated Antigen 1, AlphaPolypeptide)
- Antigen symbol:ITGAL
- Clonality:Monoclonal
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human