Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB44206-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:CD4 molecule
- Antigen symbol:CD4
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human