Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HLA-Class I Mouse Monoclonal Antibody
Anti-HLA-Class I Mouse Monoclonal Antibody
Catalog # BIRBORB44076-0.1
Supplier:  Biorbyt
Anti-HLA-Class I Mouse Monoclonal Antibody
Catalog # BIRBORB44076-0.1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-Class I
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: HLA-Class I
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human