Anti-MS4A1 Mouse Monoclonal Antibody
Catalog # BIRBORB43941-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-Domains, Subfamily A, Member 1
- Antigen symbol:MS4A1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 test
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human