Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB43831-100
 :  Biorbyt
Anti-SELP Mouse Monoclonal Antibody (PE (Phycoerythrin))
  BIRBORB43831-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Selectin P
  • Antigen symbol:
    SELP
  • Clonality:
    Monoclonal
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 test
  • Pk:
    100 Tests

 

 

Type: Primary
Antigen: SELP (selectin P (granule membrane protein 140kDa, antigen CD62))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: