Anti-HIST2H2BE Rabbit Polyclonal Antibody
BIRBORB43385-100
: Biorbyt
- Antibody type:Primary
- Antigen name:histone cluster 2, H2be
- Antigen symbol:HIST2H2BE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HIST2H2BE (histone cluster 2, H2be)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat