Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-MAP2K5 Rabbit Polyclonal Antibody
Anti-MAP2K5 Rabbit Polyclonal Antibody
  BIRBORB43353-100
 :  Biorbyt
undefined
Anti-MAP2K5 Rabbit Polyclonal Antibody
  BIRBORB43353-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase kinase 5
  • Antigen symbol:
    MAP2K5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Modification:
    phospho-Thr315
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: MAP2K5 (mitogen-activated protein kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat