Anti-IgG Mouse Monoclonal Antibody
BIRBORB43306-100
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Gel shift:Yes
- Host:Mouse
- Reactivity:Goat
- Western blot:Yes
- Storage buffer:TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
- Molecular weight:50 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:The immunogen used is goat IgG.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Goat