Anti-CFL2 Rabbit Polyclonal Antibody
Catalog # BIRBORB41820-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Cofilin 2
- Antigen symbol:CFL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1 to 127
- Storage buffer:0,1M Tris, 0,1M Glycine, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,24 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 127 of Cofilin 2.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: CFL2 (cofilin 2 (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human