Anti-14-3-3 beta Rabbit Polyclonal Antibody
BIRBORB41782-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:14-3-3 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:1 to 141
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 141 of 14-3-3 beta .
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: 14-3-3 beta (14-3-3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human