Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-EIF5A2 Rabbit Polyclonal Antibody
  BIRBORB41631-100
 :  Biorbyt
Anti-EIF5A2 Rabbit Polyclonal Antibody
  BIRBORB41631-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Eukaryotic Translation Initiation Factor 5A2
  • Antigen symbol:
    EIF5A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    89 to 153
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    0,87 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide contain a sequence corresponding to a region within amino acids 89 and 153 of Human EIF5A2.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Western Blot: 1:500-1:3000

Type: Primary
Antigen: EIF5A2 (eukaryotic translation initiation factor 5A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human