Anti-PLEKHG4 Rabbit Polyclonal Antibody
Catalog # BIRBORB41550-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Pleckstrin homology domain containing G3
- Antigen symbol:PLEKHG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:35 to 226
- Storage buffer:0,1M Tris, 0,1M Glycine, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 35 and 226 of PLEKHG4.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PLEKHG4 (pleckstrin homology domain containing, family G (with RhoGef domain) member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human