Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-RPA 70 kDa subunit Rabbit Polyclonal Antibody
  BIRBORB41485-100
 :  Biorbyt
Anti-RPA 70 kDa subunit Rabbit Polyclonal Antibody
  BIRBORB41485-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen symbol:
    RPA 70 kDa subunit
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    369 to 616
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    0,49 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 369 and 616 of RPA 70 kDa subunit.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: RPA 70 kDa subunit
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human