Anti-TRIM37 Rabbit Polyclonal Antibody
Catalog # BIRBORB41234-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 37
- Antigen symbol:TRIM37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:902 to 964
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1,02 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 902 and 964 of TRIM37.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: TRIM37 (tripartite motif containing 37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human