Anti-STOML1 Rabbit Polyclonal Antibody
Catalog # BIRBORB41047-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:stomatin (EPB72)-like 1
- Antigen symbol:STOML1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Amino acid number:106 to 338
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 106 and 338 of Human STOML1.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:3000
Type: Primary
Antigen: STOML1 (stomatin (EPB72)-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human