Anti-CACNA1S Rabbit Polyclonal Antibody
BIRBORB40951-100
: Biorbyt
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, L type, alpha 1S subunit
- Antigen symbol:CACNA1S
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:544 to 855
- Storage buffer:1XPBS, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:0,67 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 544 and 855 of CACNA1S.
- Purification:purified by antigen affinity chromatography
- Size:100 µl
- Pk:100 µl
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500
Type: Primary
Antigen: CACNA1S (calcium channel, voltage-dependent, L type, alpha 1S subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human