Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-ARAF Rabbit Polyclonal Antibody
  BIRBORB40834-100
 :  Biorbyt
Anti-ARAF Rabbit Polyclonal Antibody
  BIRBORB40834-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    v-raf murine sarcoma 3611 viral oncogene homolog
  • Antigen symbol:
    ARAF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    371 to 524
  • Storage buffer:
    0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
  • Storage temperature:
    Cool
  • Concentration:
    0,85 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein fragment contain a sequence corresponding to a region within amino acids 371 and 524 of ARAF.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: ARAF (v-raf murine sarcoma 3611 viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human