Anti-ERG Rabbit Polyclonal Antibody
Catalog # BIRBORB40768-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:v-ets avian erythroblastosis virus E26 oncogene homolog
- Antigen symbol:ERG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:395 to 457
- Storage buffer:1XPBS, 1%BSA, 20% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:4,67 mg/ml
- Shipping temperature:Ice
- Immunogen:395 - 457 of Human ERG.
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human