Anti-C-reactive protein Mouse monoclonal antibody
Catalog # BIRBORB27752-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:C-Reactive Protein
- Antigen symbol:CRP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Storage buffer:Solution in PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:E.coli-derived recombinant human C-Reactive Protein (CRP)
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: C-reactive protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human