Anti-VEGFC Mouse Monoclonal Antibody
Catalog # BIRBORB27522-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Vascular endothelial growth factor C
- Antigen symbol:VEGFC
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant human VEGF-C
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: VEGFC (vascular endothelial growth factor C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human