Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-COL1 Rabbit Polyclonal Antibody
 :  Biorbyt
 
Anti-COL1 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB27306-0.1
  • BIRBORB27306-0.5
  • Antigen symbol:
  • COL1
  • COL1
  • Antigen name:
  • collagen, type I
  • collagen, type I
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Purified collagen type I from murine skin
  • Purified collagen type I from murine skin
  • Form:
  • lyophilized
  • lyophilized
  • Purification:
  • purified
  • purified
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together