Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-COL3 Rabbit Polyclonal Antibody
Anti-COL3 Rabbit Polyclonal Antibody
Catalog # BIRBORB26849-0.5
Supplier:  Biorbyt
Anti-COL3 Rabbit Polyclonal Antibody
Catalog # BIRBORB26849-0.5
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    collagen, type III
  • Antigen symbol:
    COL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Bovine
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Collagen Type III extracted and purified from bovine skin
  • Purification:
    purified by ion exchange chromatography
  • Size:
    500 μL
  • Pk:
    0,5 mL

Specifications

About this item

Immunofluorescence: 1:20

Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine