Anti-S-100 (beta-beta) Mouse Monoclonal Antibody
Catalog # BIRBORB26771-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:S-100 (beta-beta)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:3,3 mg/ml (Sigma protein assay kit)
- Shipping temperature:Ice
- Immunogen:Purified S-100 protein from human brain
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: S-100 (beta-beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: