Anti-ITGA2 Mouse Monoclonal Antibody
Catalog # BIRBORB26534-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 2
- Antigen symbol:ITGA2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:lyophilized
- Storage buffer:PBS containing 1 mg/mL BSA.
- Storage temperature:Cool
- Concentration:0,2 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Isolated Human Platelets
- Purification:purified by Protein A chromatography
- Size:200 μg
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: ITGA2 (integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human