Anti-CD40 B Cells Mouse Monoclonal Antibody
Catalog # BIRBORB26510-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:CD40 B Cells
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized
- Storage buffer:PBS containing 1 mg/mL BSA.
- Storage temperature:Cool
- Concentration:0,2 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Anti-mu activated tonsillar human B cells.
- Size:200 μg
- Pk:0,2 mg
Specifications
About this item
Flow Cytometry: 2 ?g
Type: Primary
Antigen: CD40 B Cells
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human