Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-FTH1 Mouse Monoclonal Antibody
Anti-FTH1 Mouse Monoclonal Antibody
Catalog # BIRBORB26148-1
Supplier:  Biorbyt
Anti-FTH1 Mouse Monoclonal Antibody
Catalog # BIRBORB26148-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ferritin heavy polypeptide
  • Antigen symbol:
    FTH1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Storage buffer:
    10mM Phosphate, pH 7,4 containing 150mM Sodium chloride
  • Storage temperature:
    Cool
  • Concentration:
    4,8 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Ferritin from human spleen
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human