Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CRP Mouse monoclonal antibody
Anti-CRP Mouse monoclonal antibody
Catalog # BIRBORB25856-1
Supplier:  Biorbyt
undefined
Anti-CRP Mouse monoclonal antibody
Catalog # BIRBORB25856-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-Reactive Protein
  • Antigen symbol:
    CRP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    2,8 mg/ml (Sigma)
  • Shipping temperature:
    Ice
  • Immunogen:
    Human CRP, derived from pleural/ascetic fluid or plasma.
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: CRP (C-reactive protein, pentraxin-related)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human