Anti-SAA@ Mouse Monoclonal Antibody
Catalog # BIRBORB24701-0.5
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Serum Amyloid A1
- Antigen symbol:SAA1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Storage buffer:0,01M PBS, pH 7,2
- Storage temperature:Cool
- Concentration:1 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Highly purified recombinant human SAA
- Purification:purified by Protein G chromatography
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Western Blot: 0.1 ?g-0.5 ?g
Type: Primary
Antigen: SAA@ (serum amyloid A1 cluster)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human