Anti-TSHB Goat Polyclonal Antibody
Catalog # BIRBORB24607-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Thyroid stimulating hormone, beta
- Antigen symbol:TSHB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Storage buffer:10 mM Sodium Phosphate, pH 7,2 containing 0,15 M Sodium Chloride.
- Storage temperature:Cool
- Concentration:6,68 mg/ml (OD280, E1%=13)
- Shipping temperature:Ice
- Immunogen:Thyroid Stimulating Hormone from human pituitary glands.
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: TSHB (thyroid stimulating hormone, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human