Anti-PAPPA Mouse Monoclonal Antibody
Catalog # BIRBORB24386-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:pregnancy-associated plasma Protein A, pappalysin 1
- Antigen symbol:PAPPA
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:5,2 mg/ml (Sigma Protein Assay Kit)
- Shipping temperature:Ice
- Immunogen:Purified human PAPP-A/proMBP heterotetrameric complex (htPAPP-A).
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: PAPPA (pregnancy-associated plasma protein A, pappalysin 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human