Anti-HCV Core Antigen Mouse monoclonal antibody [clone: 7A1]
Catalog # BIRBORB24130-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:HCV Core Antigen
- Antigen symbol:HCV
- Clonality:Monoclonal
- Clone:7A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Amino acid number:80 to 120
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant HCV core antigen, a.a. 80-120
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HCV Core Antigen
Clonality: monoclonal
Clone: 7A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: