Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Toxoplasma gondii Mouse Monoclonal Antibody
Anti-Toxoplasma gondii Mouse Monoclonal Antibody
Catalog # BIRBORB24023-1
Supplier:  Biorbyt
Anti-Toxoplasma gondii Mouse Monoclonal Antibody
Catalog # BIRBORB24023-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Toxoplasma gondii
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    7,0 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Tachyzoites lysate.
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: