Anti-Hantavirus (Puumala) Mouse Monoclonal Antibody
Catalog # BIRBORB23664-0.4
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Hantavirus
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Antigen synonyms:Puumala
- Storage buffer:PBS, pH 7,2 containing 0,5% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant Puumala Nucleocapsid Protein
- Size:400 μL
- Pk:0,4 mL
Specifications
About this item
Immunochemistry: 1:50
Type: Primary
Antigen: Hantavirus (Puumala)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: