Anti-Pneumocystis carinii Mouse Monoclonal Antibody
Catalog # BIRBORB23591-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Pneumocystis carinii
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Western blot:Yes
- Form:lyophilized
- Storage buffer:PBS, pH 7,4 containing 0,5% BSA
- Molecular weight:50 kDa
- Storage temperature:Cool
- Concentration:(prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Pneumocystis carinii isolates
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunofluorescence: 1:20
Type: Primary
Antigen: Pneumocystis carinii
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: