Anti-West Nile Virus Rabbit Polyclonal Antibody
Catalog # BIRBORB23299-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:West Nile Virus
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Storage buffer:PBS
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide corresponding to 14 amino acids at the C-terminus of the West Nile Virus Envelope protein
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: West Nile Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: