Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-HBsAg Goat Polyclonal Antibody
Catalog # BIRBORB23041-1
Supplier:  Biorbyt
Anti-HBsAg Goat Polyclonal Antibody
Catalog # BIRBORB23041-1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HBsAg
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Storage buffer:
    10 mM Sodium Phosphate, pH 7,4 containing 0,15 M Sodium Chloride.
  • Storage temperature:
    Cool
  • Concentration:
    4,97 mg/ml (OD280, E1%=13)
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified HBsAg
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: HBsAg
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: