Anti-CD28 Hamster Monoclonal Antibody (FITC (Fluorescein))
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG (Hamster)
Reactivity: Mouse
Specifications
- Catalog No:
- BIRBORB22697-1
- BIRBORB22697-500
- Antigen symbol:
- CD28
- CD28
- Antigen name:
- CD28 molecule
- CD28 molecule
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mouse
- Mouse
- Host:
- Hamster
- Hamster
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Purified mouse purified mouse LN cells.
- Purified mouse purified mouse LN cells.
- Purification:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together