Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HRH2 Goat Polyclonal Antibody
Anti-HRH2 Goat Polyclonal Antibody
Catalog # BIRBORB22498-100
Supplier:  Biorbyt
undefined
Anti-HRH2 Goat Polyclonal Antibody
Catalog # BIRBORB22498-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    histamine receptor H2
  • Antigen symbol:
    HRH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,32% BSA._x000D_
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    SHNSHKTSLRLNNS
  • Purification:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HRH2 (histamine receptor H2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human