Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
BIRBORB21953-1
: Biorbyt
- Antibody type:Secondary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Peroxidase/IgG protein molar ratio (E/P) is approximately 1,7, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:A pool of purified Bence Jones kappa proteins isolated from mouse urine. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: Mouse Ig Kappa
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: