Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Platelet Factor 4Platelet Factor 4 Rabbit Polyclonal Antibody
Anti-Platelet Factor 4Platelet Factor 4 Rabbit Polyclonal Antibody
  BIRBORB21922-1
 :  Biorbyt
Anti-Platelet Factor 4Platelet Factor 4 Rabbit Polyclonal Antibody
  BIRBORB21922-1
 :  Biorbyt

 

  • Antibody type:
    Secondary
  • Antigen name:
    Platelet Factor 4
  • Antigen symbol:
    PF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Format:
    antiserum
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Secondary
Antigen: Platelet Factor 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: