Anti-IgG2a/IgG2b Goat Polyclonal Antibody
BIRBORB21763-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG2a/IgG2b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Pools of purified homogenous IgG2a and IgG2b isolated from rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: Rat IgG2Ab
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: