Anti-IgG Rabbit Polyclonal Antibody (Biotin)
BIRBORB21611-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Biotin/IgG protein molar ratio (B/P) is approximately 6,2, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG isolated from pooled cat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: IgG (Cat) (cat IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: