Anti-TAF7L Goat Polyclonal Antibody
BIRBORB20301-100
: Biorbyt
- Antibody type:Primary
- Antigen name:TAF7-like RNA polymerase II, TATA box binding Protein (TBP)-associated factor, 50kDa
- Antigen symbol:TAF7L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:40 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C Terminus
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:16000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: TAF7L (TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human